Learn More
Greiner Bio-One 25 mL Sterile Serological Pipets
Drip-free pipetting design with high optical clarity and maximum accuracy.
Brand:  Greiner Bio-One 760160
Additional Details : Weight : 3.15000kg
 View more versions of this product
Product Code. 10449943
  • € 155.00 / Pack of 200
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
Specifications
Specifications
Shape: Standard
Sterile
2/10
25 mL
Polystyrene
Yes
Individually Wrapped in plastic/plastic packaging
Serological Pipet