Learn More
STEMCELL Technologies 3 cc Syringes
For accurate dispensing of methylcellulose-based medium
Brand:  STEMCELL Technologies 28230
Additional Details : Weight : 0.26670kg
 View more versions of this product
Product Code. 17148071
  • € 59.40 / Pack of 30
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
  • Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy
Specifications
Specifications
Colony Assay
30 Syringes/Bag
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers