Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration
Optimized for a variety of applications requiring larger per well volume processing.
€ 205.00 - € 446.00
Product Code | Brand | Pore Size | Material (Membrane) | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Material (Membrane) | Volume (Metric) | Price | Quantity & Availability | |||
17114601
|
Cytiva (formerly Pall Lab)
8019 |
0.2 μm | Supor (PES) | 350 μL |
€ 320.00
Pack of 10 |
In Stock
Log in to see stock. |
|||
17194611
|
Cytiva (formerly Pall Lab)
8119 |
0.2 μm | Supor (PES) | 1 mL |
€ 205.00
Pack of 5 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
17164631
|
Cytiva (formerly Pall Lab)
8231 |
1.0 μm | Borosilicate Glass w/o Binder, Supor (PES) | 2 mL |
€ 205.00
Pack of 5 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
17134601
|
Cytiva (formerly Pall Lab)
8027 |
30 to 40 μm | Polypropylene/Polyethylene Non-woven Media | 350 μL |
€ 446.00
Pack of 10 |
In Stock
Log in to see stock. |
|||
17154601
|
Cytiva (formerly Pall Lab)
8031 |
1.0 μm | Borosilicate Glass w/o Binder, Supor (PES) | 350 mL |
€ 213.00
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
17104621
|
Cytiva (formerly Pall Lab)
8127 |
30 to 40 μm | Polypropylene/Polyethylene Non-woven Media | 1 mL |
€ 233.00
Pack of 5 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
17134621
|
Cytiva (formerly Pall Lab)
8131 |
1.0 μm | Borosilicate Glass w/o Binder, Supor (PES) | 1 mL |
€ 205.00
Pack of 5 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
17154631
|
Cytiva (formerly Pall Lab)
8227 |
30 to 40 μm | Polypropylene/Polyethylene Non-woven Media | 2 mL |
€ 247.00
Pack of 5 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Non-sterile | |
Polypropylene Housing |