Learn More
B Braun™ Aesculap™ Safety Scalpel
Reduces the risk of infections caused by cuts and stab wounds suffered during handover, take-over and disposal of scalpels.
€ 18.90 - € 38.10
Specifications
Blade Material | Carbon Steel |
---|---|
Description | Surgeon Grade |
Disposable | Yes |
Sterility | Sterile |
Includes | Protective Cap |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15313515
|
B Braun™
BA836SU |
# 36 |
€ 19.40
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15363505
|
B Braun™
BA811SU |
# 11 |
€ 19.80
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
15772577
|
B Braun™
BA815SU |
# 15 |
€ 38.10
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15782577
|
B Braun™
BA820SU |
# 20 |
€ 35.90
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15303515
|
B Braun™
BA813SU |
# 13 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Safety locking mechanism for disposal
- Single-handed operation
- Ergonomic guide for precise incision
- Ruler in cm and inches
- Transparent cap for blade protection – blade and position is always visible
- Provides optimal sharpness
Specifications
Carbon Steel | |
Yes | |
Protective Cap | |
10/Pk. |
Surgeon Grade | |
Sterile | |
Plastic |