Learn More
B Braun™ Aesculap™ Sterile Scalpel Blades #13
Made from extra-hard carbon steel with a high cutting quality.
Brand: B Braun™ BB513
Additional Details : Weight : 0.99000kg
Description
- Manufacturing techniques based on high-tech craftsmanship, know-how, experience and care
- Pointed, sharp and simply indispensable
- Excellent for dental and surgical applications
Specifications
Carbon Steel | |
# 13 | |
10/Pk. |
Dental and Surgical Applications | |
Yes |