Learn More
Alcohol Burner
Enhance your experimentation using an Alcohol Burner with wick and cap when applying heat to laboratory substances.
Brand:  N/A 2162/100
Additional Details : Weight : 0.15660kg
Product Code. 12812174
  • € 8.10 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
Enhance your experimentation using an Alcohol Burner with wick and cap when applying heat to laboratory substances.
  • With wick and cap
  • 100mL capacity
  • For burning methylated spirits during laboratory for experimentation
Order Info
CapacityMetric
Specifications
Specifications
Lamp
Cap, Wick
0.16356 kg
100 mL
1 Ea.