Learn More
ATAGO™ Automatic Digital Refractometer, Model: RX-7000i
Device to measure wider- refractive index range. The ATAGO Automatic Digital Refractometer (RX-7000i) features an intuitive touch screen technology for easy navigation and selection.
Brand:  ATAGO™ 3279
Additional Details : Weight : 15.00000kg
Product Code. 12601585
  • € 9475.00 / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Includes USB flash drive and self-diagnosis capability functions
  • Calibration with water
  • LCD digital display
  • Automatic temperature compensation
  • User scales can be inputted
  • Can be connected to printer
  • Measurement at set temperature
Four measurement mode options
  • MODE-1: Displays the measurement value once the sample reaches the target temperature.
  • MODE-2: Measures refractive index and temperature at fixed intervals and displays the estimated measurement value at the target temperature.
  • MODE-3: The thermo-module can be turn off. Without temperature control, the measurement value is displayed in 4 seconds after the START key is pressed.
  • MODE-S: Displays the measurement value once a certain level of sample stability is achieved
Specifications
Specifications
0.00 to 100.00 Brix
Refractive index (nD) : ±0.0001 Brix : ±0.1%
5°C to 75°C
1.32422 to 1.70000
Refractive index (nD) : 0.00001 (0.0001)Brix : 0.01% (0.1%)