Learn More
Bode Chemie™ Baktolan™ Cleansing Lotion
Keep skin smooth and pliable even with intensive and repeated daily use with Bode Chemie™ Baktolan™ Cleansing Lotion with Allantoin and wheat protein. The pH value of 5.5 preserves the sensitive protective acid mantle of the skin.
€ 6.76 - € 13.83
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
11336261
|
Bode Chemie™
9811373 |
100 mL | Cleansing Lotion |
€ 13.83
100mL |
In Stock
Log in to see stock. |
||||
15179786
|
Bode Chemie™
9813333 |
500 mL | Sensitive Cleansing Lotion |
€ 6.76
500mL |
In Stock
Log in to see stock. |
||||
Description
- Shelf Life:12 Months after opening
- pH Range: approximately 5.5
- Quantity: 1Pack
- Fragrance: Yes
Specifications
1 Ea. |