Learn More
BRAND™ BLAUBRAND™ AR-GLAS™ Type 3 Class AS Measuring Pipets
Manufactured from AR-GLAS™ Soda lime glass. BRAND™ Blaubrand™ AR-GLAS™ Type 3 Class AS Measuring Pipets include graduations with DE-M marking and are calibrated for total delivery. Type 3 with zero on top.
€ 71.40 - € 146.54
Product Code | Brand | Volume (Metric) | Tolerance | Graduations | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Tolerance | Graduations | Price | Quantity & Availability | |||
11888261
|
BRAND™
27712 |
5 mL | ±0.030 mL | 0.1 mL |
€ 83.40
Pack of 12 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
11878261
|
BRAND™
27711 |
5 mL | ±0.030 mL | 0.05 mL |
€ 85.80
Pack of 12 |
In Stock
Log in to see stock. |
|||
11898281
|
BRAND™
27706 |
1 mL | ±0.007 mL | 0.01 mL |
€ 71.40
Pack of 12 |
In Stock
Log in to see stock. |
|||
11858291
|
BRAND™
27705 |
0.5 mL | ±0.006 mL | 0.01 mL |
€ 90.60
Pack of 12 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
11868291
|
BRAND™
27707 |
1 mL | ±0.007 mL | 0.1 mL |
€ 71.40
Pack of 12 |
In Stock
Log in to see stock. |
|||
11878291
|
BRAND™
27708 |
2 mL | ±0.010 mL | 0.01 mL |
€ 93.00
Pack of 12 |
In Stock
Log in to see stock. |
|||
11888291
|
BRAND™
27709 |
2 mL | ±0.010 mL | 0.02 mL |
€ 71.40
Pack of 12 |
In Stock
Log in to see stock. |
|||
11808301
|
BRAND™
27715 |
25 mL | ±0.100 mL | 0.1 mL |
€ 87.60
Pack of 6 |
In Stock
Log in to see stock. |
|||
11898291
|
BRAND™
27714 |
20 mL | ±0.100 mL | 0.1 mL |
€ 87.60
Pack of 6 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
11828311
|
BRAND™
27716 |
50 mL | ±0.20 mL | 0.5 mL |
€ 146.54
Pack of 6 |
In Stock
Log in to see stock. |
|||
Description
- Manufactured from AR-GLAS™ Soda lime glass.
- Type 3 (zero on top)
- DE-M Marking graduations
- Calibrated to deliver (TD, Ex.)
- Supplied with batch number and one batch certificate per packing unit
- USP individual certificate or DAkkS calibration certificate available upon request.