Learn More
BRAND™ Borosilicate Glass Filter Adapter
Borosilicate Glass Filter Adapter suitable for use with filter crucibles.
Brand: BRAND™ 463616
328.47 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 3.80000kg
Description
- Suitable for filter crucibles
Specifications
1 D Filter Crucibles | |
Filter Adapter | |
41 mm |
Borosilicate Glass (3.3) | |
125 mm |