missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Filter Adapter

Borosilicate Glass Filter Adapter suitable for use with filter crucibles.

Brand:  BRAND™ 463732

348.77 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.



Additional Details : Weight : 5.66000kg

 View more versions of this product

Product Code. 15282459

  • € 394.00 / Pack of 10
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Suitable for filter crucibles
Specifications

Specifications

2 D Filter Crucibles
Filter Adapter
50 mm
Borosilicate Glass (3.3)
132 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers