Learn More
Erlab™ Captair by Erlab™ Benchcaps
Use these Erlab™ benchcaps with CaptairFlex ductless fume hoods.
Brand:  Erlab™ I0515000302
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 15369703
  • € 1107.70 / Each
Estimated Shipment: 03-09-2024
to see stock.
Description
Description
  • Fixed metal stand for CaptairFlex ductless fume hoods
  • Equipped with 4 cylinders to upgrade the unit
Specifications
Specifications
Benchcap
Captair
CaptairFlex Midcap M321/M321 ductless fume hoods