Learn More
Erlab™ Captair™ Mobile Ductless Fume Hood Filters
Provides personal protection while handling liquid, gaseous and solid chemicals. Erlab™ Captair™ Mobile Ductless Fume Hood Filters provide filtration of organic, acid, formadehyde, and ammonia vapors as well as fine powders.
€ 433.50 - € 1160.70
Product Code | Brand | Filter Type | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | For Use With (Application) | For Use With (Equipment) | Price | Quantity & Availability | |||
10688990
|
Erlab™
H01074201 |
BE | For Organic Vapors and Acid Vapors | Mobile Ductless Filtration Fume Hood |
€ 1022.90
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11831545
|
Erlab™
H11074201 |
BE | For Organic Vapors and Acid Vapors | Mobile Ductless Filtration Fume Hood |
€ 1022.90
Each |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12192540
|
Erlab™
H11074301 |
K | For Ammonia Vapors | Mobile Ductless Filtration Fume Hood |
€ 1160.70
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12010393
|
Erlab™
H11074101 |
AS | For Organic Vapors | Mobile Ductless Filtration Fume Hood |
€ 694.30
Each |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15109598
|
Erlab™
H01074301 |
K | For Ammonia Vapors | 834/1634 Mobile Ductless Filtration Fume Hood |
€ 1160.70
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11851545
|
Erlab™
H11074401 |
F | For Formaldehyde Vapors | Mobile Ductless Filtration Fume Hood |
€ 433.50
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11841545
|
Erlab™
H11074101 |
AS | For Organic Vapors | Mobile Ductless Filtration Fume Hood |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15341191
|
Erlab™
H01074101 |
AS | For Organic Vapors | All Captair Hoods |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Provides personal protection during handlings of liquid, gaseous and solid chemicals.Filter types available:
- GF4 AS: Oganic vapors
- GF4 BE +: Organix vapors plus acid vapors
- GF4 F: Formaldehyde vapors
- GF4 K: Ammonia vapors
- GF4 HP: HI4 HEPA filter for powders
Specifications
Replacement Filter |