Learn More
Abnova™ CEN1 (Orange) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Brand:  Abnova™ FC0182.200uL
Additional Details : Weight : 0.02000kg
Product Code. 16092755
  • € 1770.00 / 200µL
Estimated Shipment: 27-06-2024
to see stock.
Specifications
Store at -20°C in the dark.
FISH
200 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers