missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Centrifuge Tubes and Rigid Lab Plastics Zero Waste Box™
Recycle Centrifuge Tubes and Rigid Lab Plastics using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
€ 129.00 - € 223.00
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17835835
|
Terracycle™
CS129-S-NL |
Small |
€ 129.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17885735
|
Terracycle™
CS129-M-NL |
Medium |
€ 171.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17825925
|
Terracycle™
CS129-L-NL |
Large |
€ 223.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
Use this Zero Waste Box to recycle:
- Any brand and type of rigid lab plastics: centrifuge tubes, plastic bottles, trays, vials and beakers.
Approximate waste units accepted:
- Small: 200 units
- Medium: 550 units
- Large: 1000 units
Do not send in:
- Tubes and rigid lab plastics that are contaminated with bodily fluids and materials considered hazardous or dangerous
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Important:
- We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
- Please make sure that you do not mix hazardous waste with non-hazardous waste.
- As the waste producer, it is your responsibility to classify the waste.
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |