Learn More
Azpack™ Matte Vellum Paper Labels
Adhere to a variety of surfaces. Azpack™ Matte Vellum Paper Labels can be used with a thermal transfer printer to create labels.
€ 18.37 - € 28.45
Product Code | Brand | Dimensions (LxW) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Description | Price | Quantity & Availability | ||||
10491274
|
Azpack™
10491274 |
49 x 25.4 mm | Deep die cut wide edge |
€ 23.05
Pack of 500 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10193474
|
Azpack™
10193474 |
24 x 13 mm | Deep die cut wide edge |
€ 28.45
Pack of 1000 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10079972
|
Azpack™
LAB-200-071P |
64 x 25 mm | Butt cut short edge |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10204724
|
Azpack™
LAB-200-091J |
75 x 25 mm | Butt cut short edge |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Matte white vellum paper
- Thermal transfer
- Glassine finish
- Permanent adhesive backing
Specifications
White | |
Vellum |