Learn More
Hach Lange™ COD Cuvette Tubes
Standard pre-dosed, ready to use, pack of 25 COD Cuvette tubes features convenient and error-free dosing of reagents and provides precise and reliable measurements of values.
€ 132.00 - € 209.50
Specifications
Certifications/Compliance | ISO 6060-1989, DIN 38409-H41-H44 |
---|---|
Type | COD Cuvette Tube |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10714087
|
Hach Lange™
LCK314 |
15 to 150mg/L |
€ 209.50
Pack of 25 |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10415815
|
Hach Lange™
LCK1414 |
5 to 60mg/L |
€ 132.00
Pack of 25 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- COD chemical oxygen demand cuvette tube cell vial test 15 - 150mg/L O2 (pack of 25)
- For use with DR3900, DR6000, DR1900, DR2800, DR3800, and DR5000 instruments
- Cuvette test, (tube test / cell test / vial test)
- Photometric, chemical oxygen demand (COD)
- 15 - 150mg/L O2 measurement range
- Dichromate method according to ISO 6060-1989 standard and DIN 38409-H41-H44
- Standard pre-dosed, ready to use, pack of 25
Specifications
ISO 6060-1989, DIN 38409-H41-H44 | |
COD Cuvette Tube |