Learn More
Witeg™ Color Comparing Cylinders Acc. to Nessler
For the comparison of color and turbidity of solutions.
Brand:  Witeg™ 2 150 050
Additional Details : Weight : 0.80000kg
Product Code. 11843855
  • € 27.50 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Tall form
  • Capacity of 50 mL
  • Manufactured from borosilicate glass
  • With optically plane bottom
Specifications
Specifications
50 mL
180 mm
Comparison of color, turbidity
Borosilicate Glass
24 mm
50 mL