missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 1/4 in. Flow Size

Detach these fittings with a simple press of the thumb latch. Cole-Parmer™ CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Brand:  CPC™ 31311-68

Additional Details : Weight : 0.80000kg

Product Code. 11770199

  • € 26.85 / Pack of 5
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Flow Rate: Cv∽5.5 (79.25) max.

  

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials
  • O-Ring: Silicone Seal USP Class VI

  

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

 

Specifications

Specifications

Quick-Disconnect Fitting
Autoclavable
19.1 mm
N/A
Purple
USP Class VI, ADCF
Polycarbonate
31.5 mm
-40°C
121° C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers