Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ B0496731
Additional Details : Weight : 0.10000kg
Product Code. 10578270
  • € 215.00 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • With inscription label
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
500 mL
Duran Glass, Borosilicate Glass
Pressure Equalizing Pipe Interior
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers