Learn More
Memmert™ DAkkS Factory Calibration for IN450
DAkkS factory calibration for a freely selectable temperature for Memmert Incubators.
Brand:  Memmert™ IN450E39696
Additional Details : Weight : 0.01200kg
Product Code. 17507735
  • € 2095.00 / Each
Estimated Shipment: 25-06-2024
to see stock.
Description
Description
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
IN450 Incubator
DAkkS Factory Calibration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers