Learn More
Esculape Laboratoire™ Diphoterine Spray Neutralizing Agent
Use Esculape Laboratoire™ Diphoterine Spray Neutralizing Agent. This hypertonic solution attracts the chemical mixture product that has penetrated the tissues and neutralizes it;by osmotic effect.
Brand:  Esculape Laboratoire™ 8205210
Additional Details : Weight : 0.10000kg Transport : UN number : 1950 Chem class : 2.2
Product Code. 11873061
  • € 114.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Sterile polyvalent
  • Hypertonic solution
  • Used to wash face and body
  • 100mL
Specifications
Specifications
170 x 45 mm
170 mm