Learn More
B Braun™ Dispensing Pin Mini-Spike Filter
Brand: B Braun™ 4550234
Additional Details : Weight : 0.45900kg
Specifications
50 Pack | |
Mini-Spike Filter | |
For filtered injection or filtered withdrawal of fluid from vials or semi-rigid containers with 0.45μm air-filter and 5μm fluid filter | |
Blue |