Learn More
B Braun™ Dispensing Pin Mini-Spike Filter
Brand:  B Braun™ 4550234
Additional Details : Weight : 0.45900kg
Product Code. 10396192
  • € 126.00 / Pack of 50
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Specifications
50 Pack
Mini-Spike Filter
For filtered injection or filtered withdrawal of fluid from vials or semi-rigid containers with 0.45μm air-filter and 5μm fluid filter
Blue
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers