Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
Brand: 3M™ 7000006981
Additional Details : Weight : 0.25000kg
Description
3M Particulate Respirator P1 P1 for mechanically generated particles grinding, sanding, crushing and sawing Not TGA Approved Not allowed for Clinical Use
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.Specifications
European Community Directive 89/686/EEC, EN 149:2001 A1:2009, CE | |
Single-use | |
Particulate Respirator | |
Protection against dust and mist | |
8822 | |
Dusts, mists and Metal fumes |
Twin strap | |
FFP2 NR D | |
Yes | |
Yes | |
10/Pk. | |
Convex |