Learn More
Semadeni™ Disposable Polystyrene Mouth Spatulas
With absolutely smooth, edge- and burr-free design.
Brand:  Semadeni™ 3337
Additional Details : Weight : 0.99000kg
Product Code. 10742865
  • € 21.00 / Pack of 100
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Not sterilized
  • White in color
  • Strong and viscoplastic
  • Physiologically harmless
  • Provided in dust-proof packaging
Specifications
Specifications
Mouth Spatula
Polystyrene
100/Pk.
150 mm
Oral examinations
18 mm