Learn More
BTX™ ECM™ 2001+ HV Output Cable
ECM™ 2001+ HV Output Cable for use with BTX™ ECM™ 2001+.
€ 420.00 - € 650.00
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery |
---|---|
For Use With (Equipment) | ECM™ 2001+ |
Product Type | Cables and Adapters |
Includes | Cable Only |
Quantity | 1 Ea. |
Product Code | Brand | Length (Metric) Cable | Length (English) Cable | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Cable | Length (English) Cable | Price | Quantity & Availability | ||||
15801947
|
BTX™
452057 |
1.5 m | 5 ft. |
€ 420.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15811947
|
BTX™
452058 |
3 m | 10 ft. |
€ 650.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- For connection to electroporation generators
Specifications
In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery | |
Cables and Adapters | |
1 Ea. | |
Output Cable |
ECM™ 2001+ | |
Cable Only | |
Non-sterile | |
AC Sine Wave, DC Square Wave |