Learn More
Merck SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames
Mini and midi blot holding frames for SNAP i.d. 2.0 Protein Detection Systems
€ 158.00 - € 362.00
Specifications
Includes | Clear lids, easy locking latch, umbrella valve |
---|---|
Stackable | Yes |
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
13479569
|
Merck
SNAP2FRMD01 |
Midi Blot Holding Frames | 1/Pk. |
€ 158.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
13248319
|
Merck
SNAP2FRMN01 |
Mini Blot Holding Frames | 1/Pk. |
€ 197.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
13258319
|
Merck
SNAP2FRMN02 |
Mini Blot Holding Frames | 2/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13489569
|
Merck
SNAP2FRMD02 |
Midi Blot Holding Frames | 2/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
For use with EMD Millipore SNAP i.d. 2.0 Protein Detection System (Cat. No. SNAP2MM)
Specifications
Clear lids, easy locking latch, umbrella valve | |
Yes |