Learn More
Eppendorf Eppendorf twin.tec™ PCR plate 96, semi-skirted
Made from polypropylene, Eppendorf twin.tec PCR Plates LoBind enable you to recover nearly 100 % of your DNA/RNA molecules, and as there’s no surface coating, there’s no risk of sample contamination. Batch-tested and certified free from DNA, DNase, RNase and PCR inhibitors, they are perfect for PCR.
Brand:  Eppendorf 0030129695
Additional Details : Weight : 0.42000kg
Product Code. 16070582
  • € 84.55 / Pack of 10
Estimated Shipment: 14-06-2024
to see stock.
Description
Description
Eppendorf twin.tec™ PCR plate 96, semi-skirted
Specifications
Specifications
10
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers