Learn More
Thermo Scientific™ Eutech™ Salt Bridge for pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1
Thermo Scientific™ offers Eutech™ Salt Bridge as a replacement part for Eutech™ pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1.
Brand:  Thermo Scientific™ 15X403101
Additional Details : Weight : 0.10000kg
Product Code. 11553865
  • € 165.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
Eutech™ Salt Bridge is used for pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1. The electrodes are designed for continuous use in industrial process applications.
Features:
  • A replaceable heavy-duty quad junction salt bridge for extended life in severe applications.
Specifications
Specifications
pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1
Replacement Part