Learn More
Lenz Laborglasinstrumente™ Evaporating Flask
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ B0260551
Additional Details : Weight : 0.44100kg
Product Code. 10264120
  • € 76.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • With inscription label
  • Available in a range of capacities
  • Center neck
Specifications
Specifications
2000 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers