Learn More
UVP Filter Pen-Ray shortwave absorbs visible light while transmitting ultraviolet at 254nm UVP
Brand:  UVP 98-0017-01
Additional Details : Weight : 0.10000kg
Product Code. 12803828
  • € 140.00 / Each
Estimated Shipment: 18-07-2024