Learn More
Fisherbrand™ Vortex Mixer Accessory, Cup Head
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.03000kg
Product Code. 15776949
  • € 36.80 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Designed for mixing one tube at a time
Designed for mixing one tube at a time.
Specifications
Specifications
Replacement Cup Head
Vortex mixer