Learn More
Camlab™ Formazine Turbidity Standard 4000NTU
The Barnant 246142 formazin standard solution has a 4000 NTU turbidity and is used for calibration of turbidimeters.
Brand: Camlab™ 246142
Additional Details : CAS : 503-27-5 Weight : 0.12200kg
Description
The Barnant 246142 formazin standard solution has a 4000 NTU turbidity and is used for calibration of turbidimeters.
- Formazin Turbidity Standard, 4000 NTU, 100 mL
- Stablcal turbidity standard for all turbidimeters
- The Barnant 246142 formazin standard solution has a 4000 NTU turbidity and is used for calibration of turbidimeters.
- The bottle has a 100mL volume
- The bottle has a concentration of 4000 NTU
Specifications
Formazine Turbidity Standard | |
4000 NTU | |
100 mL |
Safety and Handling
- StablCal Formazin Turbidity Standard, 4000 NTU
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methenamine; hexamethylenetetramine