Learn More
Poulten & Graf GmbH FORTUNA™ OPTIMAT™ Automatic Dispensing Station
Poulten & Graf GmbH FORTUNA™ OPTIMAT™ Automatic Dispensing Station is suitable for one dispensing pump.
Brand:  Poulten & Graf GmbH 150 0003
Additional Details : Weight : 0.10000kg
Product Code. 10319690
  • € 24300.00 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Suitable for one dispensing pump
  • 8 pump sizes available up to 300mL
  • With RS232 connection interface
  • Pump and tubes are not included
Specifications
Specifications
50 Hz
No
2 - 300mL
230V
RS 232 connection interface
Automatic Dispensing Station without pumps