missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brema™ Ice Makers GB 1555 HC Granular Ice Flaker
Self-contained granular ice flaker
Brand: Brema™ Ice Makers PBA24
Additional Details : Weight : 94.00000kg
Description
- Granular ice - 80-82% dry
- 1 L (water) - 1kg (ice) - only Air cooled
- AISI 304 SS Scotch Brite finish
- Tropical class +43°C
- Adjustable Stainless Steel feet
- Removable and cleanable air filter
- AISI 304 SS evaporator
- Air or water cooling system
- Easy cleanable internal rounded surfaces
- HCFC-Free insulated storage
- Dust-proof water basin
- RoHS Free
- Disappearing insulated door
- Electromechanical operation
- ON-OFF rocker switch
- Automatic water charge
- Standard and special voltages
- HFC Free
Specifications
1020 mm | |
690 mm | |
10 A | |
153 kg per day | |
+21°C |
Air | |
220-240 V (50 Hz) | |
738 mm | |
55 kg | |
700 W |