Learn More
Fisherbrand™ Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Pipette with the highest precision. Fisherbrand™ one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
€ 5.10 - € 23.20
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15227935
|
Fisherbrand™
|
1 mL |
€ 5.10
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15237935
|
Fisherbrand™
|
2 mL |
€ 5.75
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15247935
|
Fisherbrand™
|
5 mL |
€ 8.50
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15257935
|
Fisherbrand™
|
10 mL |
€ 8.25
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15267935
|
Fisherbrand™
|
25 mL |
€ 10.50
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15277935
|
Fisherbrand™
|
50 mL |
€ 14.30
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15287935
|
Fisherbrand™
|
100 mL |
€ 23.20
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
Pipette with the highest precision. Fisherbrand one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
- Mark one pipet for precision measurment
- Type one pipet with zero mark at the top
- Soda lime glass
- Batch Certificate of Compliance is downloadable
Specifications
Soda Lime Glass | |
Yes | |
Pipet |