Learn More
Witeg™ Glass Petri Dish
Culture your cells and see them clearly in a Witeg™ Glass Petri Dish, with accompanying lid.
€ 3.10 - € 860.00
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
10796595
|
Witeg™
5 830 006 |
120 mm | 20 mm |
€ 15.38
Each |
In Stock
Log in to see stock. |
||||
10194434
|
Witeg™
5 830 010 |
200 mm | 50 mm |
€ 183.00
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10092944
|
Witeg™
5 830 007 |
150 mm | 25 mm |
€ 178.00
Pack of 18 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10296534
|
Witeg™
5 830 090 |
90 mm | 15 mm |
€ 4.60
Each |
In Stock
Log in to see stock. |
||||
10431555
|
Witeg™
5 830 003 |
80 mm | 15 mm |
€ 3.10
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10387844
|
Witeg™
5 830 009 |
200 mm | 30 mm |
€ 860.00
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10276154
|
Witeg™
5 830 008 |
180 mm | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Free from bubbles and other imperfections
- With lid
- Glass
Specifications
Glass | |
Round | |
Petri Dish |