Learn More
Gosselin™ Polypropylene Buckets with Handle
Excellent option for general storage in the laboratory. Gosselin™ Polypropylene Buckets with Handle are manufactured from white-colored polypropylene plastic.
Brand: Gosselin™ JT38-01
Additional Details : Weight : 12.50000kg
Description
- Nonsterile
- Convenient carrying handle
- Closure with tamper-evident seal
Specifications
Polypropylene | |
EC N°1935/2004 | |
Snap cap, Handles and Lids |
Yes | |
White | |
75 Pk. |