missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
€ 11.60 - € 282.87
Specifications
Color | Natural |
---|---|
Material | LDPE |
Type | Tapered Closure |
For Use With (Application) | For use as plugs in bores or internal thread, or as caps for external thread |
Product Code | Brand | Diameter (Metric) Outside | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Length (Metric) | Price | Quantity & Availability | ||||
10718235
|
Poeppelmann™
60014680000 |
159.5 mm | 33.5 mm |
€ 93.90
Pack of 25 |
In Stock
Log in to see stock. |
||||
10033654
|
Poeppelmann™
60009900000 |
110 mm | 17.5 mm |
€ 96.44
Pack of 50 |
In Stock
Log in to see stock. |
||||
10023514
|
Poeppelmann™
60011400000 |
125 mm | 17.2 mm |
€ 115.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10217764
|
Poeppelmann™
60018360000 |
199.5 mm | 338 mm |
€ 130.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10747805
|
Poeppelmann™
60001600000 |
20 mm | 8.8 mm |
€ 14.95
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10297404
|
Poeppelmann™
60029900000 |
315 mm | 22.5 mm |
€ 282.87
Pack of 25 |
In Stock
Log in to see stock. |
||||
10338724
|
Poeppelmann™
60003450000 |
41 mm | 14 mm |
€ 31.11
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10517204
|
Poeppelmann™
60002120000 |
26 mm | 14 mm |
€ 15.85
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
10472565
|
Poeppelmann™
60020600000 |
216 mm | 23.5 mm |
€ 135.19
Pack of 25 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||||
10378714
|
Poeppelmann™
60001980000 |
24 mm | 8.8 mm |
€ 11.60
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
10145114
|
Poeppelmann™
60002300000 |
28 mm | 13.8 mm |
€ 12.75
Pack of 100 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10115364
|
Poeppelmann™
60004980000 |
56 mm | 13.5 mm |
€ 41.50
Pack of 100 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
10266844
|
Poeppelmann™
60005550000 |
61 mm | 13.5 mm |
€ 48.40
Pack of 100 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10535485
|
Poeppelmann™
60006600000 |
72 mm | 13 mm |
€ 74.85
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10195314
|
Poeppelmann™
60007400000 |
85 mm | 17 mm |
€ 172.34
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Natural | |
LDPE | |
Tapered Closure | |
For use as plugs in bores or internal thread, or as caps for external thread |