Learn More
B Braun™ Hand Pump with Reflux Valve
For use with 1000 mL bottles.
Brand: B Braun™ 3908381
Additional Details : Weight : 0.01800kg
Description
- Round head
- Reflux valve
- Single use
- Dosage of approx. 2 mL
- No measuring device
Specifications
Hand Pump | |
Dispensing liquids | |
Reflux valve |
White | |
1000 mL bottles |