Learn More
Rotronic™ HC2-C05 Model Stainless Steel Miniature Cable Probe
Brand:  Rotronic™ HC2C05
Additional Details : Weight : 0.13000kg
Product Code. 11900675
  • € 1005.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Specifications
Miniature Cable Probe
3.2 to 5 VDC
1 Each
Stainless Steel
4.5 mA