Learn More
Brady™ Honeywell Enhanced Xenon 1952H HD Bluetooth Scanner
The wireless area-imaging scanner Xenon 1952H HD delivers superior bar code scanning and digital image capture.
Brand:  Brady™ 306803
Additional Details : Weight : 1.50000kg
Product Code. 17282304
  • € 1440.00 / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Xenon 1952H HD incorporates a revolutionary decoding architecture and a custom sensor, enabling extended depth of field, faster reading, and improved scanning performance on poor quality bar codes
  • From high density linear to 2D bar codes found directly on the screen of a mobile device, Xenon 1952H HD decodes virtually all bar codes with ease. Xenon 1952H HD incorporates a Bluetooth Class 2, v2.1 radio, enabling unrestricted movement up to 10 m from the base, and a battery that provides up to 14 hours of battery life
  • For added convenience, a paging system helps locate misplaced scanners
Specifications
Specifications
High Density Imager Sensor
Lithium-ion Battery, 500 mA (at 5 V DC)
Bluetooth, USB