Learn More
Boekel Scientific™ Hybridization Glass Bottles for Boekel Little Shot Oven
For use with Boekel Little Shot Oven, these glass hybridization bottles fit the standard carousel and help maintain the perfect temperature and mixing for all applications.
€ 146.00 - € 212.00
Specifications
Color | Blue |
---|---|
Diameter (English) Inner | 1.37 in. |
Diameter (Metric) Inner | 35 mm |
Material | Glass |
Product Type | Hybridization Bottle |
Product Code | Brand | Length (Metric) | Length (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Length (English) | Price | Quantity & Availability | ||||
10465743
|
Boekel Scientific™
502-0100 |
100 mm | 3.93 in. |
€ 146.00
Pack of 2 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
10784994
|
Boekel Scientific™
502-0150 |
150 mm | 5.9 in. |
€ 154.00
Pack of 2 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10475743
|
Boekel Scientific™
502-0225 |
225 mm | 8.85 in. |
€ 212.00
Pack of 2 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10345393
|
Boekel Scientific™
502-0300 |
300 mm | 11.8 in. |
€ 200.00
Pack of 2 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
- Pack of 2 Bottles
- Comes with Blue Caps
- For Use with Standard Carousel
Specifications
Blue | |
35 mm | |
Hybridization Bottle |
1.37 in. | |
Glass | |
Blue caps and silicone O-ring |