Learn More
Gilson™ Hydrophobic Membrane Filter
Brand:  Gilson™ F1077416
Additional Details : Weight : 0.02000kg
Product Code. 15672197
  • € 43.10 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • This hydrophobic, non-sterile membrane filter (diameter 60 mm, 0.45 μm) is for use with the Gilson Safe Aspiration Station.
Specifications
Specifications
Hydrophobic Membrane Filter
Gilson™ Safe Aspiration Station
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers