missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Industrial Safety Equipment Zero Waste Box™
Recycle Industrial Safety Equipment using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
€ 131.00 - € 231.00
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17835795
|
Terracycle™
CS199-S-NL |
Small |
€ 131.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17875695
|
Terracycle™
CS199-M-NL |
Medium |
€ 176.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17865795
|
Terracycle™
CS199-L-NL |
Large |
€ 231.00
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
Use this Zero Waste Box to recycle:
- Industrial safety helmets
- Safety bump caps
- Hi-vis jackets
- Heavy-duty gloves
- Ear defenders Protective eyewear (goggles)
Do not send in:
- Fabric work gloves with an anti-slip layer
- Suede and leather gloves
- Disposable face masks
- Safety footwear
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass, and medical waste
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |