missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Inlet Liner Shimadzu GC
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Marke: Trajan Scientific and Medical™ 09229201
Weitere Details : Weight : 0.10000kg
Beschreibung
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Shimazdu instruments
Spezifikation
Straight | |
1 Ea. | |
1.5 mm | |
Shimadzu 2010 GC Instruments |
GC Inlet Liner | |
95 mm | |
3.1 mm | |
Non-Sintered LVI with Wool |