Learn More
B Braun™ FEP Safety IV Catheters, Introcan
Use B Braun™ FEP Safety IV Catheters, Introcan™. From insertion to catheter deployment to needle removal, protection is provided by an automatically activated system that cannot be bypassed.
€ 83.15 - € 148.00
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
12301113
|
B Braun™
4254562-01 |
18 G |
€ 148.00
Pack of 50 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10453202
|
B Braun™
4252071B |
24 G |
€ 83.95
Pack of 50 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174713
|
B Braun™
4252098B |
22 G |
€ 83.15
Pack of 50 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Color-coded labels for easy gauge size identification
- Back cut bevel
- FEP for firmer construction for arterial access and special procedures
- DEHP- and latex-free
Specifications
Introcan |