Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Filtering Flask
Provides mechanical strength under vacuum application
€ 166.00
Specifications
Diameter (Metric) Inner Tubing 0.93 cm
Material Borosilicate Glass
View More Specs
Products 1
Produktcode Marke Capacity (Metric) Preis Menge & Verfügbarkeit  
Produktcode Marke Capacity (Metric) Preis Menge & Verfügbarkeit  
15374626
missing translation for 'viewDocuments' Promotion Details
DWK Life Sciences
617500-0003
250 mL
€ 166.00
Each
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 12-11-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Outer joint: 24/40
  • Serrated hose connection: No. 2
Spezifikation
Spezifikation
0.93 cm
Borosilicate Glass
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate