Learn More
DWK Life Sciences Kimble™ Kontes™ Neoprene Gasket for NMR Tube Washer
Replacement gasket for utility washers.
Brand: DWK Life Sciences 459951-0000
Additional Details : Weight : 0.10000kg
Description
- Utility washers are used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items
- Unit is supplied with one gasket, a tube washer, an Erlenmeyer flask and a 3/8 in. hose connection
- Replacement gaskets, Case of 12
Specifications
Neoprene Gasket | |
Neoprene | |
For NMR utility washer | |
Neoprene Gasket |