Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™ Cordless Motor: Replacement Motor Adapter
Replacement parts for Kontes Pellet Pestle Cordless Motor
Brand:  DWK Life Sciences 749541-0000
Additional Details : Weight : 1.00000kg
Product Code. 11824055
  • € 15.15 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
Replacement parts for Kontes Pellet Pestle Cordless Motor
For use with Kontes Pellet Pestle Cordless Motor (Cat. No. K749540-0000)
Specifications
Specifications
Rubber
Replacement Motor Adapter Sleeve
Pellet Pestle Cordless Motor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers